Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

thioredoxin domain containing 3 (spermatozoa), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018478 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-478 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-478 Supplier Abnova Corporation Supplier No. H00051314D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human TXNDC3 protein.

TXNDC3 encodes a group II thioredoxin protein, composed of a thioredoxin domain and 3 NDP kinase domains, with testis-specific expression.[supplied by OMIM

Sequence: MASKKREVQLQTVINNQSLWDEMLQNKGLTVIDVYQAWCGPCRAMQPLFRKLKNELNEDEILHFAVAEADNIVTLQPFRDKCEPVFLFSVNGKIIEKIQGANAPLVNKKVINLIDEERKIAAGEMARPQYPEIPLVDSDSEVSEESPCESVQELYSIAIIKPDAVISKKVLEIKRKITKAGFIIEAEHKTVLTEEQVVNFYSRIADQRDFEEFVSFMTSGLSYILVVSQGSKHNPPSEETEPQTDTEPNERSEDQPEVEAQVTPGMMKNKQDSLQEYLERQHLAQLCDIEEDAANVAKFMDAFFPDFKKMKSMKLEKTLALLRPNLFHERKDDVLRIIKDEDFKILEQRQVVLSEKEAQALCKEYENEDYFNKLIENMTSGPSLALVLLRDNGLQYWKQLLGPRTVEEAIEYFPESLCAQFAMDSLPVNQLYGSDSLETAEREIQHFFPLQSTLGLIKPHATSEQREQILKIVKEAGFDLTQVKKMFLTPEQTEKIYPKVTGKDFYKDLLEMLSVGPSMVMILTKWNAVAEWRRLMGPTDPEEAKLLSPDSIRAQFGISKLKNIVHGASNAYEAKEVVNRLFEDPEEN

Specifications

Antigen thioredoxin domain containing 3 (spermatozoa)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human TXNDC3 protein.
Formulation PBS with no preservative; pH 7.4
Gene TXNDC3
Gene Accession No. NM_016616.2
Gene Alias CILD6/NME8/SPTRX2
Gene Symbols TXNDC3
Host Species Rabbit
Immunogen TXNDC3 (NP_057700.2, 1 a.a. ∼ 588 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51314
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.