Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyk2 (pY1054/pY1055) Mouse anti-Human, Unlabeled, Clone: I114-617, BD

Catalog No. BDB558394 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558394 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558394 Supplier BD Biosciences Supplier No. 558394
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tyk2 is a widely expressed protein tyrosine kinase (PTK) of 1187 amino acids (135kDa). The N-terminal kinase-related domain of Tyk2 shows 54% identity to the kinase-related domain of the JAK1 kinase. Surprisingly, the catalytic region of JAK1 shares more sequence identity to Tyk2 than it does with JAK2 kinase. The similarity of structure between Tyk2 and JAK1 suggests that both may be involved in related phosphorylation cascades. Tyk2 plays a critical role in the interferon (IFN)-a/b response while JAK1 shows involvement in both the IFN- α/β and IFN-γ signal transduction pathways. JAK2, however, appears to be required only in the IFN-g response. It has been suggested that a portion of the large extracatalytic domain of Tyk2 could interact with components of the IFN a/b receptor. The I114-617 monoclonal antibody recognizes the phosphorylated Y1054 and Y1055 in the tyrosine kinase domain of Tyk2.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Tyk2 (pY1054/pY1055)
Applications Western Blot
Classification Monoclonal
Clone I114-617
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated peptide of the region including the tyrosines 1054 and 1055 of human Tyk2
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.