Learn More
Description
Thymidylate synthase catalyzes the methylation of deoxyuridylate to deoxythymidylate using 5,10-methylenetetrahydrofolate (methylene-THF) as a cofactor. This function maintains the dTMP (thymidine-5-prime monophosphate) pool critical for DNA replication and repair. The enzyme has been of interest as a target for cancer chemotherapeutic agents. It is considered to be the primary site of action for 5-fluorouracil, 5-fluoro-2-prime-deoxyuridine, and some folate analogs. Expression of this gene and that of a naturally occuring antisense transcript rTSalpha (GeneID:55556) vary inversely when cell-growth progresses from late-log to plateau phase. [provided by RefSeq
Specifications
Specifications
| Antigen | thymidylate synthetase |
| Applications | ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot |
| Classification | Monoclonal |
| Clone | 3A1 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant TYMS. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | TYMS |
| Gene Accession No. | BC013919 |
| Gene Alias | HsT422/MGC88736/TMS/TS/TSase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.