Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYRO3 protein tyrosine kinase, Mouse, Clone: 4F6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant TYRO3.
Supplier: Abnova Corporation H00007301M05
Description
Sequence: VKLTVSQGQPVRLNCSVEGMEEPDIQWVKDGAVVQNLDQLYIPVSEQHWIGFLSLKSVERSDAGRYWCQVEDGGETEISQPVWLTVEGVPFFTVEPKDLAVSpecifications
TYRO3 protein tyrosine kinase | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC049368 | |
TYRO3 | |
TYRO3 (AAH49368, 50 a.a. ∼ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Cell Adhesion | |
7301 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA, KnockDown, Western Blot | |
4F6 | |
Mouse monoclonal antibody raised against a partial recombinant TYRO3. | |
TYRO3 | |
BYK/Brt/Dtk/FLJ16467/RSE/Sky/Tif | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction