Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYRO3 protein tyrosine kinase, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018345 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-345 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-345 Supplier Abnova Corporation Supplier No. H00007301D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human TYRO3 protein.

Sequence: MALRRSMGRPGLPPLPLPPPPRLGLLLAALASLLLPESAAAGLKLMGAPVKLTVSQGQPVKLNCSVEGMEEPDIQWVKDGAVVQNLDQLYIPVSEQHWIGFLSLKSVERSDAGRYWCQVEDGGETEISQPVWLTVEGVPFFTVEPKDLAVPPNAPFQLSCEAVGPPEPVTIVWWRGTTKIGGPAPSPSVLNVTGVTQSTMFSCEAHNLKGLASSRTATVHLQALPAAPFNITVTKLSSSNASVAWMPGADGRALLQSCTVQVTQAPGGWEVLAVVVPVPPFTCLLRDLVPATNYSLRVRCANALGPSPYADWVPFQTKGLAPASAPQNLHAIRTDSGLILEWEEVIPEAPLEGPLGPYKLSWVQDNGTQDELTVEGTRANLTGWDPQKDLIVRVCVSNAVGCGPWSQPLVVSSHDRAGQQGPPHSRTSWVPVVLGVLTALVTAAALALILLRKRRKETRFGQAFDSVMARGEPAVHFRAARSFNRERPERIEATLDSLGISDELKEKLEDVLIPEQQFTLGRMLGKGEFGSVREAQLKQEDGSFVKVAVKMLKADIIASSDIEEFLREAACMKEFDHPHVAKLVGVSLRSRAKGRLPIPMVILPFMKHGDLHAFLLASRIGENPFNLPLQTLIRFMVDIACGMEYLSSRNFIHRDLAARNCMLAEDMTVCVADFGLSRKIYSGDYYRQGCASKLPVKWLALESLADNLYTVQSDVWAFGVTMWEIMTRGQTPYAGIENAEIYNYLIGGNRLKQPPECMEDVYDLMYQCWSADPKQRPSFTCLRMELENILGQLSVLSASQDPLYINIERAEEPTAGGSLELPGRDQPYSGAGDGSGMGAVGGTPSDCRYILTPGGLAEQPGQAEHQPESPLNETQRLLLLQQGLLPHSSC

Specifications

Antigen TYRO3 protein tyrosine kinase
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human TYRO3 protein.
Formulation PBS with no preservative; pH 7.4
Gene TYRO3
Gene Accession No. NM_006293.2
Gene Alias BYK/Brt/Dtk/FLJ16467/RSE/Sky/Tif
Gene Symbols TYRO3
Host Species Rabbit
Immunogen TYRO3 (NP_006284.2, 1 a.a. ∼ 890 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Adhesion
Primary or Secondary Primary
Gene ID (Entrez) 7301
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.