Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tyrosine Hydroxylase Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5986
Description
This Anti-Tyrosine Hydroxylase Antibody is validated for use in immunohistochemistry for the detection of Tyrosine Hydroxylase.
Specifications
Tyrosine Hydroxylase | |
Polyclonal | |
Rabbit serum. Lyophilized. Reconstitute with 50μL of sterile distilled water. Centrifuge to remove any insoluble material. Contains no preservative. | |
Rabbit | |
50 μL | |
NM_000360.3; NM_199292.2; NM_199293.2 | |
Lyophilized: −20-−70°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. Avoid repeated freeze/thaw cycles. |
Immunohistochemistry | |
Unconjugated | |
TH; TYH; EC 1.14.16.2 | |
Synthetic peptide corresponding to amino acids 32-47 of rat Tyrosine Hydroxylase. | |
Primary | |
Rat | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction