Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Tyrosine Hydroxylase Alexa Fluor 488, Clone: LNC1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MAB318AF488
Description
Specifically detects Tyrosine Hydroxylase Clone: LNC1 in Chicken, Frog, Human, Mouse, Rat, Zebrafish samples, and it is validated for Immunocytochemistry
Tyrosine 3-monooxygenase (EC 1.14.16.2; UniProt P07101; also known as Dystonia 14, TH, Tyrosine 3-hydroxylase) is encoded by the TH (also known as DYT14, DYT5b, TYH) gene (Gene ID 7054) in human. Tyrosine hydroxylase plays an important role in the physiology of adrenergic neurons. It is the first and rate-limiting enzyme involved in the biosynthesis of the catecholamines Dopamine and Norepinephrine from tyrosine. TH is, therefore, a useful marker for dopaminergic and noradrenergic neurons. TH enzymatic activity requires ferrous ions as cofactors and is believed to be regulated by phosphorylation. Six isoforms of human TH exist as a result of alternative splicing (UniProt P07101-1 through P07101-6).
Specifications
Tyrosine Hydroxylase | |
Monoclonal | |
Alexa Fluor 488 | |
P07101 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Recognizes an epitope on the outside of the regulatory N-terminus of tyrosine hydroxylase. Does not react with dopamine-beta-hydroxylase, phenylalanine hydroxylase, trytophan hydroxylase, dehydropteridine reductase, sepiapterin reductase, or phenethanolamine-N-methyl transferase (PNMT) by Western blotting using the unconjugated antibody. | |
2°C to 8°C for one year from date of shipment | |
IgG1 κ |
Immunocytochemistry | |
LNC1 | |
Purified mouse antibody conjugate in PBS with 15mg/mL BSA and 0.1 % sodium azide. | |
TH, TYH | |
Tyrosine Hydroxylase purified from PC12 cells. | |
100 μL | |
Neuroscience | |
NM_000360.3 | |
Chicken, Frog, Human, Mouse, Rat, Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction