Learn More
Tyrosine Hydroxylase Mouse anti-Bovine, Chicken, Quail, Rat, Clone: 2/40/15, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Tyrosine Hydroxylase |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 2/40/15 |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal IgG2a in buffer containing 0.02MPhosphate buffer, 0.25MNaCl, pH 7.6 with 0.1% sodium azide. |
| Gene Accession No. | NM_000360.3 |
| Gene Symbols | TH, TYH, EC 1.14.16.2 |
| Host Species | Mouse |
| Immunogen | Purified tyrosine hydroxylase (EC 1.14.16.2) from a rat pheochromocytoma (2). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.