Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrosine Hydroxylase Mouse, Unlabeled, Clone: 45, BD

Catalog No. BDB612300 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612300 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612300 Supplier BD Biosciences Supplier No. 612300

Mouse Monoclonal Antibody

Dopamine and its metabolic products norepinephrine and epinephrine are catecholamine neurotransmitters whose function is essential for the execution of normal neural processes in the CNS and PNS. Tyrosine hydroxylase (TH) is a non-heme iron, tetrahydrobiopterin-dependent enzyme that catalzyes the conversion of tyrosine to L-dihydroxyphenylalanine (L-DOPA). This is the rate-limiting step in the biosynthesis of catecholamines. Both the development of Parkinson's disease and other neurodegenerative diseases result from loss of the ability to synthesize catecholamines. Decreases in the activity of TH have been implicated in these diseases. Nitration of TH at Tyrosine 423 has been associated with temporary loss of enzymatic activity, and TH nitration occurs in response to the Parkinsonian toxin MPTP, and following exposure to peroxynitrite. These findings implicate nitration as a potential mode of down-regulation of TH activity during neurodegenerative disease. Thus, TH is an essential enzyme for catecholamine synthesis, which is required for normal neuronal function.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Tyrosine Hydroxylase
Applications Western Blot
Classification Monoclonal
Clone 45
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat Tyrosine Hydroxylase aa. 18-133
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.