Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrosine Hydroxylase Rabbit anti-Rat, Mouse, Human, Avian, Polyclonal, MilliporeSigma™

Catalog No. AB5935MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5935MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5935MI Supplier MilliporeSigma Supplier No. AB5935
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Tyrosine Hydroxylase Antibody, phosphoSer 40 detects level of Tyrosine Hydroxylase and has been published and validated for use in immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tyrosine Hydroxylase, phosphoSer 40
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal liquid in buffer containing 10mM Hepes, pH 7.5, 150mM NaCl, 100μg/mL BSA, and 50% glycerol.
Gene Symbols TH; TYH; EC 1.14.16.2
Host Species Rabbit
Immunogen Synthetic peptide from the phosphoSer40 of rat Tyrosine Hydroxylase.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000360.3
Target Species Avian, Human, Mouse, Rat
Content And Storage -20°C in undiluted aliquotes for up to 6 months, Avoid Freeze/Thaw Cycles
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.