Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Tyrosine Hydroxylase, Polyclonal

Catalog No. 657012100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
65-701-2100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-701-2100UL Supplier MilliporeSigma™ Supplier No. 657012100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Tyrosine Hydroxylase Clone: in Many Species samples, and it is validated for Dot Blot, Immunofluorescence, Immunohistochemistry (Frozen), Immuno Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Tyrosine Hydroxylase
Applications Dot Blot, Immunoblot, Immunoblot, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Unconjugated
Formulation In 150mM NaCl, 10mM HEPES, 50% glycerol, 0.01% BSA, pH 7.5.
Host Species Rabbit
Immunogen purified, SDS-denatured rat pheochromocytoma tyrosine hydroxylase
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽60kDa tyrosine hydroxylase protein in okadaic acid-treated PC12 cells.
Target Species Multi-species
Content And Storage ≤ -70°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.