Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human u-Plasminogen Activator (uPA)/ Urokinase Catalytic Domain Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB1310500 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1310500 500 μg
MAB1310SP 25 μg
MAB1310100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB1310500 Supplier R&D Systems Supplier No. MAB1310500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

u-Plasminogen Activator/Urokinase Monoclonal specifically detects u-Plasminogen Activator/Urokinase in Human samples. It is validated for ELISA Capture (Matched Antibody Pair)∣Immunoprecipitation∣Western Blot.

Specifications

Antigen u-Plasminogen Activator/Urokinase
Applications ELISA Capture (Matched Antibody Pair), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 204212
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P00749
Gene Alias ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, PLAU, uPA, u-PA, uPlasminogen Activator, U-plasminogen activator, urinary, Urokinase, urokinase-type plasminogen activator
Gene Symbols PLAU
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human u-Plasminogen Activator (uPA)/Urokinase Catalytic Domain Ser21-Leu431 Accession # P00749
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5328
Test Specificity Detects human u-Plasminogen Activator (uPA)/Urokinase Catalytic Domain in direct ELISAs and Western blots. In Western blots under reducing conditions, it reacts with the catalytic domain (B chain) only. In Western blots under non-reducing conditions, it reacts with the B chain and disulfide bond-linked B and A chains. In direct ELISAs, no cross-reactivity with recombinant human (rh) Factor X, rhHGFA, rhKLK-3, -4, -5, -6, -8, -10, -11, rhThrombin, or recombinant mouse WIF-1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.