Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

U1-70K Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 061297MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-129-7MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-129-7MI Supplier MilliporeSigma Supplier No. 061297
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-U1-70K Antibody is a Rabbit Polyclonal Antibody for detection of U1-70K also known as small nuclear ribonucleoprotein 70kDa (U1) and has been validated in western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen U1-70K
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols U1RNP; U1-70K; U170K; RNPU1Z; SNRP70; U1AP1; RPU1; snRNP70; SNRNP70; U1AP
Host Species Rabbit
Immunogen Recombinant protein corresponding to C-terminal domain of human U1-70K.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_003080.2
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.