Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ubiquitin associated and SH3 domain containing, A, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018494 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-494 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-494 Supplier Abnova Corporation Supplier No. H00053347D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human UBASH3A protein.

Sequence: MAAGETQLYAKVSNKLKGRSSPSLLEPLLAMGFPVHTALKALAATGRKTAEEALAWLHDHCNDPSLDDPIPQEYALFLCPTGPLLEKLQEFWRESKRQCAKNRAHEVFPHVTLCDFFTCEDQKVECLYEALKRAGDRLLGSFPTAVPLALHSSISYLGFFVSGSPADVIREFAMTFATEASLLADCSVKPCTKQLHLTLAHKFYPHHQRTLEQLARAIPLGHSCQWTAALYSRDMRFVHYQTLRALFQYKPQNVDELTLSPGDYIFVDPTQQDEASEGWVIGISQRTGCRGFLPENYTDRASESDTWVKHRMYTFSLATDLNSRKDGEASSRCSGEFLPQTARSLSSLQALQATVARKSVLVVRHGERVDQIFGKAWLQQCSTPDGNYYRPDLNFPCSLPRRSRGIKDFENDPPLSSCGIFQSRIAGMFEDCLVGKVTITATLIMASRHPA

Specifications

Antigen ubiquitin associated and SH3 domain containing, A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human UBASH3A protein.
Formulation PBS with no preservative; pH 7.4
Gene UBASH3A
Gene Accession No. BC028138.1
Gene Alias CLIP4/STS-2/TULA
Gene Symbols UBASH3A
Host Species Rabbit
Immunogen UBASH3A (AAH28138.1, 1 a.a. ∼ 451 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53347
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.