Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubc9 Mouse, Unlabeled, Clone: 50, BD

Catalog No. BDB610748 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610748 50 μg
BDB610749 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610748 Supplier BD Biosciences Supplier No. 610748

Mouse Monoclonal Antibody

Progression of the mammalian cell cycle is primarily regulated by phosphorylation/dephosphorylation and synthesis/degradation of many key proteins. Ubiquitin, a soluble protein of 76 amino acids, is enzymatically attached to an e-NH2-Lys in a target protein. Ubiquitination is a hallmark for rapid protein degradation of the target protein in the proteosome (a cytoplasmic complex of proteases). Human homologs of the yeast ubiquitin-conjugating enzymes (Ubc) have been reported, including Ubc9. Ubc9 is 158 amino acids with an apparent molecular weight of 18kDa. Although ubiquitously expressed, the highest levels of Ubc9 are found in testis and thymus. Ubc9 was localized to the synaptonemal complex in male mouse sex chromosomes. Furthermore, Ubc9 interacts with the recombination protein Rad51, thus suggesting an important role for Ubc9 during meiosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubc9
Applications Western Blot
Classification Monoclonal
Clone 50
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Ubc9 aa. 26-156
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.