Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UbcH6 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB3858MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3858MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3858MI Supplier MilliporeSigma Supplier No. AB3858

Rabbit Polyclonal Antibody

Anti-UbcH6 Antibody is an antibody against UbcH6 for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UbcH6
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Protein A purified immunoglobulin. Liquid in PBS, pH 7.4.
Gene Symbols UBE2E1; UbcH6; UBCH6; EC 6.3.2.19
Host Species Rabbit
Immunogen Full length, highly purified, UbcH6 protein.
Purification Method Affinity Purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_003341.3; NM_182666.1
Target Species Human
Content And Storage 2-8°C for up to 1 month, −70°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.