Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UbcH7 Mouse, Unlabeled, Clone: 20, BD

Catalog No. BDB610852 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610852 50 μg
BDB610853 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610852 Supplier BD Biosciences Supplier No. 610852

Mouse Monoclonal Antibody

Progression of the mammalian cell cycle is regulated mainly by phosphorylation/dephosphorylation and synthesis/degradation of many key proteins. Ubiquitin, a soluble protein of 76 amino acids, is enzymatically attached to an ε -NH2-Lys in a target protein. Ubiquitination is a hallmark for rapid protein degradation of the target protein in the proteosome-a cytoplasmic complex of proteases. Several human homologs of the yeast ubiquitin-conjugating enzymes (Ubc) have been identified and found involved in critical processes such as cell cycle progression and apoptosis. UbcH7 was characterized as an E6-AP-interacting protein required for the ubiquitination of p53. E6-AP is the ubiquitin ligase used as bait in the search for interacting proteins required for the proper ubiquitination of cellular targets. UbcH7 interacts with the C-terminal region of E6-AP named the hect domain. Therefore, UbcH7 is another protein, like Ubc9, involved in the degradation and regulation of critical proteins in the cell cycle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UbcH7
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 20
Concentration 250μg/mL
Conjugate Unconjugated
Description Ubiquitin Conjugating Enzyme H7
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human UbcH7 aa. 15-133
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.