Learn More
ubiquitin-conjugating enzyme E2E 3 (UBC4/5 homolog, yeast), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human UBE2E3 protein.
Supplier: Abnova Corporation H00010477B01
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. The encoded protein shares 100% sequence identity with the mouse and rat counterparts, which indicates that this enzyme is highly conserved in eukaryotes. Two alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Sequence: MSSDRQRSDDESPSTSSGSSDADQRDPAAPEPEEQEERKPSATQQKKNTKLSSKTTAKLSTSAKRIQKELAEITLDPPPNCSAGPKGDNIYEWRSTILGPPGSVYEGGVFFLDITFSSDYPFKPPKVTFRTRIYHCNINSQGVICLDILKDNWSPALTISKVLLSICSLLTDCNPADPLVGSIATQYLTNRAEHDRIARQWTKRYATSpecifications
ubiquitin-conjugating enzyme E2E 3 (UBC4/5 homolog, yeast) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human UBE2E3 protein. | |
UBE2E3 | |
UBCH9/UbcM2 | |
Mouse | |
50 μL | |
Ubiquitin/Proteasome | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_006357 | |
UBE2E3 | |
UBE2E3 (NP_006348, 1 a.a. ∼ 207 a.a) full-length human protein. | |
RUO | |
Yes | |
10477 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.