Learn More
UBE2G1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human UBE2G1 peptide using ARM Technology.
Supplier: Abnova Corporation H00007326K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family and catalyzes the covalent attachment of ubiquitin to other proteins. The protein may be involved in degradation of muscle-specific proteins. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
UBE2G1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
UBE2G1 | |
UBE2G1 | |
A synthetic peptide of human UBE2G1 | |
RUO | |
7326 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
ubiquitin-conjugating enzyme E2G 1 (UBC7 homolog, yeast) | |
UBE2G1 | |
E217K, UBC7, UBE2G | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.