Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2K, Mouse, Clone: 1D10, Abnova™

Catalog No. 89123156 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-156 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-156 Supplier Abnova Corporation Supplier No. H00003093M13
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant UBE2K.

The protein encoded by this gene belongs to the ubiquitin-conjugating enzyme family. This protein interacts with RING finger proteins, and it can ubiquitinate huntingtin, the gene product for Huntington's disease. Known functions for this protein include a role in aggregate formation of expanded polyglutamine proteins and the suppression of apoptosis in polyglutamine diseases, a role in the dislocation of newly synthesized MHC class I heavy chains from the endoplasmic reticulum, and involvement in foam cell formation. Multiple transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MLAMSPPAADSYPPPPPPLRPPPPPPPLPPPPPPSAIQT*

Specifications

Antigen UBE2K
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant UBE2K.
Formulation PBS with no preservative; pH 7.4
Gene UBE2K
Gene Accession No. BC006419
Gene Alias DKFZp564C1216/DKFZp686J24237/E2-25K/HIP2/HYPG/LIG
Gene Symbols UBE2K
Host Species Mouse
Immunogen UBE2K (-, 1 a.a. ∼ 40 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3093
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.