Learn More
UBE2V2, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human UBE2V2 peptide using ARM Technology.
Supplier: Abnova Corporation H00007336K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Ubiquitin-conjugating enzyme E2 variant proteins constitute a distinct subfamily within the E2 protein family. They have sequence similarity to other ubiquitin-conjugating enzymes but lack the conserved cysteine residue that is critical for the catalytic activity of E2s. The protein encoded by this gene also shares homology with ubiquitin-conjugating enzyme E2 variant 1 and yeast MMS2 gene product. It may be involved in the differentiation of monocytes and enterocytes. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
UBE2V2 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
UBE2V2 | |
UBE2V2 | |
A synthetic peptide of human UBE2V2 | |
RUO | |
7336 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
ubiquitin-conjugating enzyme E2 variant 2 | |
UBE2V2 | |
DDVIT1, DDVit-1, EDAF-1, EDPF-1, EDPF1, MMS2, UEV-2, UEV2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.