Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquilin 2 Mouse anti-Human, Mouse, Clone: 6H9, MilliporeSigma™

Catalog No. MABN763MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN763MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN763MI Supplier MilliporeSigma Supplier No. MABN763
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Ubiquilin-2 using this mouse monoclonal antibody, Anti-Ubiquilin 2 Antibody, clone 6H9 validated for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquilin 2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 6H9
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ other in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_038472
Gene Symbols UBQLN2, N4BP4, PLIC2
Host Species Mouse
Immunogen Recombinant protein corresponding to human Ubiquilin 2.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.