Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquitin Rabbit anti-Bovine, Human, Vertebrates, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07375
Description
This Anti-Ubiquitin Antibody is validated for use in western blotting for the detection of Ubiquitin.
Specifications
Ubiquitin | |
Polyclonal | |
Protein A Purified immunoglobulin in PBS, 0.1% sodium azide, and 30% glycerol. | |
Rabbit | |
Protein A purfied | |
Primary | |
Bovine, Human, Vertebrates | |
Purified |
Western Blot | |
Unconjugated | |
RPS27A; CEP80; ubiquitin; ubiquitin-CEP80; UBCEP1; Ubiquitin.; UBA52; UBCEP2; UBA80; UBC; UBB; HUBCEP80 | |
full-length ubiquitin isolated from bovine erythrocytes. | |
200 μg | |
NM_002954.3 | |
−20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction