Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin Rabbit anti-Human, Mouse, Rat, Rabbit, Clone: Apu3, MilliporeSigma™

Catalog No. 051308MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-130-8MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-130-8MI Supplier MilliporeSigma Supplier No. 051308
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-Ubiquitin Antibody, Lys63-Specifimmunocytochemistry, clone Apu3 is a high quality Rabbit Monoclonal Antibody for the detection of Ubiquitin and has been validated in Inhibition Assay (PIA), Immunocytochemistry (immunocytochemistry), Immunoprecipitation (IP), Immunohistochemistry (immunohistochemistry) and Western Blotting (WB).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquitin
Applications Functional Assay, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone Apu3
Conjugate Unconjugated
Formulation Protein A purified recombinant Rabbit Monoclonal antibody that consists of a humanized fAb fragment fused to a rabbit Fc tail produced in a CHO recombinant antibody system. Supplied in 0.1M Tris-Glycine (pH7.4) 150mM NaCl, 0.05% sodium azide.
Host Species Rabbit
Immunogen Immunogen used was human di-ubiquitin linked at Lys63.
Purification Method Protein A purfied
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rabbit, Rat
Content And Storage 2-8°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.