Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Ubiquitin Clone: FK2,

Catalog No. ST1200100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1200100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1200100UG Supplier MilliporeSigma™ Supplier No. ST1200100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Ubiquitin Clone: FK2 in samples, and it is validated for ELISA, Immunofluorescence, Immunoprecipitation, Immuno Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquitin
Applications ELISA, Immunoblot, Immunoblot, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone FK2
Conjugate Unconjugated
Formulation In PBS.
Host Species Mouse
Immunogen unpurified polyubiquitinylated lysozyme
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes K29-, K48-, and K63-linked polyubiquitinylated and monoubiquitinylated proteins, but not free ubiquitin, in Raji cells.
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.