Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin Mouse anti-Human, Vertebrates, Yeast (S. cerevisiae), Clone: P4D1-A11, MilliporeSigma™

Catalog No. 05944MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-944-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-944-MI Supplier MilliporeSigma Supplier No. 05944
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Ubiquitin Antibody, clone P4D1-A11 is an antibody against Ubiquitin for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquitin
Applications Western Blot
Classification Monoclonal
Clone P4D1-A11
Conjugate Unconjugated
Formulation 100μg of protein G purified mouse IgG in 100μL of 70% storage buffer (0.014M phosphate buffer, pH 7.6, 0.175M NaCl, 0.07% sodium azide) and 30% glycerol.
Gene Accession No. NM_018955
Gene Symbols RPS27A, CEP80, ubiquitin, ubiquitin-CEP80, UBCEP1, Ubiquitin., UBA52, UBCEP2, UBA80, UBC, UBB, HUBCEP80
Host Species Mouse
Immunogen total bovine ubiquitin
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, S. cerevisiae, Vertebrates, Yeast
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.