Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Ubiquitin, Clone: Ubi-1 (042691GS), ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB1510/DEL

Description
Electron Microscopy, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting

Specifications
Ubiquitin | |
Unconjugated | |
Purified ubiquitin (bovine RBC) covalently coupled to KLH using glutaraldehyde. | |
RUO | |
Primary | |
Ascites |
Monoclonal | |
Mouse | |
100 μL | |
Neuroscience, Protein Trafficking | |
Human, Mouse, Rat | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction