Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin Rabbit anti-Human, AlexaFluor™647, Clone: Apu3, MilliporeSigma™

Catalog No. FCMAB288A6 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB288A6 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB288A6 Supplier MilliporeSigma Supplier No. FCMAB288A6
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

This Milli-Mark™ Anti-Ubiquitin Antibody, Lys63-Specific-Alexa Fluor™647, clone Apu3 is validated for use in FC for the detection of Ubiquitin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquitin
Applications Flow Cytometry
Classification Polyclonal
Clone Apu3
Conjugate Alexa Fluor 647
Formulation Purified recombinant rabbit monoclonal IgG1 conjugated to Alexa Fluor 647 in PBS with 0.1% sodium azide and 15mg/mL BSA.
Host Species Rabbit
Immunogen Immunogen used was di-ubiquitin linked at Lys63.
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C protected from light for up to 6 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.