Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Ubiquitin, Polyclonal

Catalog No. 66209950UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
66-209-950UL 50 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66-209-950UL Supplier MilliporeSigma™ Supplier No. 66209950UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Ubiquitin Clone: in Bovine, Canine, Chicken, Drosophila, E. coli, Frog, Guinea Pig, Hamster, Human, Monkey, Mouse, Rabbit, Rat, Sheep, Yeast samples, and it is validated for Immunohistochemistry, Immunoprecipitation

Ubiquitin is a highly conserved protein that functions in intracellular regulation. The ubiquitin system is the major pathway for selective protein degradation and is responsible for the degradation of abnormal and short-lived proteins. In the first step, ubiquitin is activated in an ATP-dependent reaction by ubiquitin-activating enzyme (E1). Activated ubiquitin is transferred to a ubiquitin conjugating enzyme (E2), then transferred to a protein substrate with the aid of ubiquitin-protein ligase (E3). An isopeptide linkage is formed between the C-terminus of the ubiquitin and an internal lysine residue of the substrate protein. Degradation requires the presence of a multi-ubiquitin chain on a substrate, which targets the protein for degradation by the 26S proteasome. Ubiquitin molecules are then released and recycled.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquitin
Applications Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Immunogen bovine erytherocyte ubiquitin conjugated to KLH
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽8.6kDa unconjugated and various forms of conjugated ubiquitin.
Target Species Bovine, Canine, Chicken, Drosophila, E. coli, Frog, Guinea Pig, Hamster, Human, Monkey, Mouse, Rabbit, Rat, Sheep, Yeast
Content And Storage −20°C
Form Undiluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.