Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitinylated proteins Mouse anti-All, Clone: FK2, MilliporeSigma™

Catalog No. 04263MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-263-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-263-MI Supplier MilliporeSigma Supplier No. 04263
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Ubiquitinylated proteins Antibody, clone FK2 has been published and validated for use in ELISA, Immunofluorescence (IF), Immunoprecipitation (IP) and Western Blotting (WB) for the detection of Ubiquitinylated proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquitinylated proteins
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone FK2
Conjugate Unconjugated
Formulation 100μL of purified immunoglobulin in PBS containing 0.1% sodium azide. Dilute to working strength with phosphate buffered saline pH 7.2-7.4.
Host Species Mouse
Immunogen Poly-ubiquitinylated-lysozyme.
Quantity 100 μL
Primary or Secondary Primary
Target Species All species
Content And Storage −20°C for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.