Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ubiquityl-Histone H2A Mouse anti-Human, Rat, Mouse, Monkey, Amphibian, Clone: E6C5

Catalog No. 05678MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-678-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-678-MI Supplier MilliporeSigma™ Supplier No. 05678
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Use Anti-ubiquityl-Histone H2A Antibody, clone E6C5 (Mouse Monoclonal Antibody) has been published and validated for use in Chimmunoprecipitation, immunocytochemistry, western blotting to detect ubiquityl-Histone H2A also known as H2AUb, Histone H2A (ubiquityl), histone H2A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ubiquityl-Histone H2A
Applications ChIP Assay, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone E6C5
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgM in buffer containing PBS with 0.05% sodium azide before the addition of glycerol to 30%. Liquid at −20deg;C.
Gene Accession No. NP_003508
Gene Symbols HIST1H2AA, H2AFR, bA317E16.2
Host Species Mouse
Immunogen AMA (human epithelial amnion) cell residual nuclear pellet portions.
Purification Method Ammonium sulfate precipitation
Quantity 200 μL
Primary or Secondary Primary
Target Species Amphibian, Human, Monkey, Mouse, Rat
Content And Storage −20°C for up to 12 months.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.