Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquityl Histone H2A.X (Lys119) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB10029MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10029MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10029MI Supplier MilliporeSigma Supplier No. AB10029
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Ubiquityl Histone H2A.X (Lys119) Antibody is a rabbit polyclonal antibody for detection of Ubiquityl Histone H2A.X (Lys119) also known as H2AXK119Ub, Histone H2A.X Ubiquityl K119) and has been validated in western blot, immunoprecipitation, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ubiquityl Histone H2A.X (Lys119)
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Unpurified rabbit polyclonal containing serum with 0.05% sodium azide.
Gene Symbols H2AFX; H2A/X; H2A.X; H2AX; H2a/x
Host Species Rabbit
Immunogen Linear peptide containing ubiquitylated Lys119 of human Histone H2A.X.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_002096.1.
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.