Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UBP1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP147214
Description
UBP1 Polyclonal specifically detects UBP1 in Human, Mouse, Rat, Canine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
UBP1 | |
Polyclonal | |
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:2000 | |
NP_055332.3 | |
UBP1 | |
Peptide with sequence C-PNHQGDGASQTSGE corresponding to internal region according to NP_001121632.1. | |
0.1 mg | |
Primary | |
Reported variants represent identical protein: NP_001121633.1, NP_055332.3. | |
Store at -20C. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
DKFZp686L1745, LBP1, LBP-1a, LBP1A, LBP1B, LBP-1B, Transcription factor LBP-1, upstream binding protein 1 (LBP-1a), upstream-binding protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
7342 | |
Human, Mouse, Rat, Canine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction