Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-UBXD1, Polyclonal

Catalog No. AB10033MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AB10033MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10033MI Supplier MilliporeSigma™ Supplier No. AB10033

Rabbit Polyclonal Antibody

Specifically detects UBXD1 Clone: in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Western Blotting

UBX domain-containing protein 1 is a component of a complex required to couple deglycosylation and proteasome-mediated degradation of misfolded proteins in the endoplasmic reticulum that are retrotranslocated in the cytosol. UBX domain-containing protein 1 is also involved in ubiquitin-proteasome systems.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBXD1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Accession No. Q9BZV1
Gene Symbols UBXN6, UBXD1, UBXDC2
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the entire protein of human UBXD1.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Trafficking
Primary or Secondary Primary
Gene ID (Entrez) NP_079517
Test Specificity This antibody recognizes UBXD1.
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C for one year
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.