Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fas associated factor family member 2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89007247 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-247 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-247 Supplier Abnova Corporation Supplier No. H00023197B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human UBXD8 protein.

The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients. [provided by RefSeq

Sequence: MAAPEERDLTQEQTEKLLQFQDLTGIESMDQCRHTLEQHNWNIEAAVQDRLNEQEGVPSVFNPPPSRPLQVNTADHRIYSYVVSRPQPRGLLGWGYYLIMLPFRFTYYTILDIFRFALRFIRPDPRSRVTDPVGDIVSFMHSFEEKYGRAHPVFYQGTYSQALNDAKRELRFLLVYLHGDDHQDSDEFCRNTLCAPEVISLINTRMLFWACSTNKPEGYRVSQALRENTYPFLAMIMLKDRRMTVVGRLEGLIQPDDLINQLTFIMDANQTYLVSERLEREERNQTQVLRQQQDEAYLASLRADQEKERKKREERERKRRKEEEVQQQKLAEERRRQNLQEEKERKLECLPPEPSPDDPESVKIIFKLPNDSRVERRFHFSQSLTVIHDFLFSLKESPEKFQIEANFPRRVLPCIPSEEWPNPPTLQEAGLSHTEVLFVQDLTDE

Specifications

Antigen Fas associated factor family member 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human UBXD8 protein.
Formulation No additive
Gene FAF2
Gene Accession No. NM_014613.1
Gene Alias ETEA/KIAA0887/UBXD8/UBXN3B
Gene Symbols FAF2
Host Species Mouse
Immunogen UBXD8 (NP_055428.1, 1 a.a. ∼ 445 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 23197
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.