Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

uridine-cytidine kinase 1-like 1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89007779 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-007-779 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-007-779 Supplier Abnova Corporation Supplier No. H00054963B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human UCKL1 protein.

Sequence: MAAPPARADADPSPTSPPTARDTPGRQAEKSETACEDRSNAESLDRLLPPVGTGRSPRKRTTSQCKSEPPLLRTSKRTIYTAGRPPWYNEHGTQSKEAFAIGLGGGSASGKTTVARMIIEALDVPWVVLLSMDSFYKVLTEQQQEQAAHNNFNFDHPDAFDFDLIISTLKKLKQGKSVKVPIYDFTTHSRKKDWKTLYGANVIIFEGIMAFADKTLLELLDMKIFVDTDSDIRLVRRLRRDISERGRDIEGVIKQYNKFVKPSFDQYIQPTMRLADIVVPRGSGNTVAIDLIVQHVHSQLEEGCAGLGTPVPPAAPDAERPEEHAAGTGHAHHHQVRAHLGTGRPRARAPAQLLAPQGQGDQSRRVHLLLQETDAAAHRARALLPALSGLRRTDPAGAGLCGQVLCGEADHRCVHSARR

Specifications

Antigen uridine-cytidine kinase 1-like 1
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human UCKL1 protein.
Formulation No additive
Gene UCKL1
Gene Accession No. BC033078.1
Gene Alias UCK1-LIKE/UCK1L/URKL1
Gene Symbols UCKL1
Host Species Mouse
Immunogen UCKL1 (AAH33078.1, 1 a.a. ∼ 419 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Kinases
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 54963
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.