Learn More
UCN3, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human UCN3 peptide using ARM Technology.
Supplier: Abnova Corporation H00114131K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene is a member of the sauvagine/corticotropin-releasing factor/urotensin I family. It is structurally related to the corticotropin-releasing factor (CRF) gene and the encoded product is an endogenous ligand for CRF type 2 receptors. In the brain it may be responsible for the effects of stress on appetite. In spite of the gene family name similarity, the product of this gene has no sequence similarity to urotensin II. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
UCN3 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
UCN3 | |
UCN3 | |
A synthetic peptide of human UCN3 | |
RUO | |
114131 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
urocortin 3 (stresscopin) | |
UCN3 | |
MGC119002, SCP, SPC, UCNIII | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.