Learn More
uncoupling protein 1 (mitochondrial, proton carrier), Mouse, Clone: 4B7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant UCP1.
Supplier: Abnova Corporation H00007350M03
Description
Mitochondrial uncoupling proteins (UCP) are members of the family of mitochondrial anion carrier proteins (MACP). UCPs separate oxidative phosphorylation from ATP synthesis with energy dissipated as heat, also referred to as the mitochondrial proton leak. UCPs facilitate the transfer of anions from the inner to the outer mitochondrial membrane and the return transfer of protons from the outer to the inner mitochondrial membrane. They also reduce the mitochondrial membrane potential in mammalian cells. Tissue specificity occurs for the different UCPs and the exact methods of how UCPs transfer H+/OH- are not known. UCPs contain the three homologous protein domains of MACPs. This gene is expressed only in brown adipose tissue, a specialized tissue which functions to produce heat. [provided by RefSeq]
Sequence: PVDVVKTRFINSPPGQYKSVPNCAMKVFTNEGPTAFSpecifications
uncoupling protein 1 (mitochondrial, proton carrier) | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_021833 | |
UCP1 | |
UCP1 (NP_068605, 232 a.a. ∼ 267 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
50 μg | |
Primary | |
Human, Mouse | |
Antibody |
ELISA, Western Blot | |
4B7 | |
Mouse monoclonal antibody raised against a partial recombinant UCP1. | |
UCP1 | |
SLC25A7/UCP | |
Mouse | |
Affinity Purified | |
RUO | |
7350 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.