Learn More
Description
UCRC is a subunit of mitochondrial complex III (ubiquinol-cytochrome c reductase; EC 1.10.2.2), which forms the middle segment of the respiratory chain of the inner mitochondrial membrane (Schagger et al., 1995 [PubMed 8592474]).[supplied by OMIM
Specifications
Specifications
| Antigen | UCRC |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 2B5 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full-length recombinant UCRC. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | UCRC |
| Gene Accession No. | BC005402 |
| Gene Alias | HSPC051/HSPC119/HSPC151 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.