Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ubiquitin-like with PHD and ring finger domains 1, Mouse, Clone: 3A11, Abnova™

Catalog No. 89014304 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-304 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-304 Supplier Abnova Corporation Supplier No. H00029128M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant UHRF1.

This gene encodes a member of a subfamily of RING-finger type E3 ubiquitin ligases. The protein binds to specific DNA sequences, and recruits a histone deacetylase to regulate gene expression. Its expression peaks at late G1 phase and continues during G2 and M phases of the cell cycle. It plays a major role in the G1/S transition by regulating topoisomerase IIalpha and retinoblastoma gene expression, and functions in the p53-dependent DNA damage checkpoint. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: WNEVLASLKDRPASGSPFQLFLSKVEETFQCICCQELVFRPITTVCQHNVCKDCLDRSFRAQVFSCPACRYDLGRSYAMQVNQPLQTVLNQLFPGYGNGR

Specifications

Antigen ubiquitin-like with PHD and ring finger domains 1
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3A11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant UHRF1.
Formulation PBS with no preservative; pH 7.4
Gene UHRF1
Gene Accession No. NM_013282
Gene Alias FLJ21925/ICBP90/MGC138707/Np95/RNF106/hNP95
Gene Symbols UHRF1
Host Species Mouse
Immunogen UHRF1 (NP_037414, 694 a.a. ∼ 793 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Gene ID (Entrez) 29128
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.