Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ubiquitin-like with PHD and ring finger domains 2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008480 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-480 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-480 Supplier Abnova Corporation Supplier No. H00115426B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human UHRF2 protein.

This gene encodes a nuclear protein which is involved in cell-cycle regulation. The encoded protein is a ubiquitin-ligase capable of ubiquinating PCNP (PEST-containing nuclear protein), and together they may play a role in tumorigenesis. [provided by RefSeq

Sequence: MWIQVRTIDGSKTCTIEDVSRKATIEELRERVWALFDVRPECQRLFYRGKQLENGYTLFDYDVGLNDIIQLLVRPDPDHLPGTSTQIEAKPCSNSPPKVKKAPRVGPSNQPSTSARARLIDPGFGIYKVNELVDARDVGLGAWFEAHIHSVTRASDGQSRGKTPLKNGSSCKRTNGNIKHKSKENTNKLDSVPSTSNSDCVAADEDVIYHIQYDEYPESGTLEMNVKDLRPRARTILKWNELNVGDVVMVNYNVESPGQRGFWFDAEITTLKTISRTKKELRVKIFMGGSEGTLNDCKIISVDEIFKIERPGAHPLSFADGKFLRRNDTECDLCGGDPEKKCHSCSCRVCGGKHEPNMQLLCDECNVAYHIYCLNPPLDKVPEEEYWYCPSCKTDSSEVVKAGERLKMSKKKAKMQSASTESRRDWGRGMACVGRTRECTIVPSNHYGPIPGIPVGSTWRFRVQVSEAGVHRPHVGGIHGRSNDGAYSLVLAGGFADEVLTEL

Specifications

Antigen ubiquitin-like with PHD and ring finger domains 2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human UHRF2 protein.
Formulation No additive
Gene UHRF2
Gene Accession No. BC028397.2
Gene Alias DKFZp434B0920/DKFZp686G0837/MGC33463/NIRF/RNF107/URF2
Gene Symbols UHRF2
Host Species Mouse
Immunogen UHRF2 (AAH28397.1, 1 a.a. ∼ 503 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 115426
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.