Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ULK2, Mouse, Clone: 2A12, Abnova™

Catalog No. 89002753 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-753 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-753 Supplier Abnova Corporation Supplier No. H00009706M05
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ULK2.

This gene encodes a protein that is similar to a serine/threonine kinase in C. elegans which is involved in axonal elongation. The structure of this protein is similar to the C. elegans protein in that both proteins have an N-terminal kinase domain, a central proline/serine rich (PS) domain, and a C-terminal (C) domain. The gene is located within the Smith-Magenis syndrome region on chromosome 17. Alternatively spliced transcript variants encoding the same protein have been identified. [provided by RefSeq

Sequence: FLRTRTTSVGPSNSGGSLCAMSGRVCVGSPPGPGFGSSPPGAEAAPSLRYVPYGASPPSLEGLITFEAPELPEETLMEREHTDTLRHLNVMLMFTECVLDL

Specifications

Antigen ULK2,
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2A12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ULK2.
Formulation PBS with no preservative; pH 7.4
Gene ULK2
Gene Accession No. BC034988
Gene Alias KIAA0623/Unc51.2
Gene Symbols ULK2
Host Species Mouse
Immunogen ULK2 (AAH34988, 743 a.a. ∼ 843 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9706
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.