Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

unc-13 homolog D (C. elegans), Mouse, Clone: 1E11, Abnova™

Catalog No. 89001405 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-405 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-405 Supplier Abnova Corporation Supplier No. H00201294M06
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant UNC13D.

This gene encodes a protein that is a member of the UNC13 family, containing similar domain structure as other family members but lacking an N-terminal phorbol ester-binding C1 domain present in other Munc13 proteins. The protein appears to play a role in vesicle maturation during exocytosis and is involved in regulation of cytolytic granules secretion. Mutations in this gene are associated with familial hemophagocytic lymphohistiocytosis type 3, a genetically heterogeneous, rare autosomal recessive disorder. [provided by RefSeq

Sequence: ATLLSHPQQRPPFLRQAIKIRRRRVRDLQDPPPQMAPEIQPPSHHFSPEQRALLYEDALYTVLHRLGHPEPNHVTEASELLRYLQEAFHVEPEEHQQTL

Specifications

Antigen unc-13 homolog D (C. elegans)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant UNC13D.
Formulation PBS with no preservative; pH 7.4
Gene UNC13D
Gene Accession No. NM_199242
Gene Alias FHL3/HLH3/HPLH3/Munc13-4
Gene Symbols UNC13D
Host Species Mouse
Immunogen UNC13D (NP_954712.1, 2 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 201294
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.