Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
unc-45 homolog B (C. elegans), Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human UNC45B protein.
Supplier: Abnova Corporation H00146862B01P
Description
UNC45B plays a role in myoblast fusion and sarcomere organization (Price et al., 2002 [PubMed 12356907]).[supplied by OMIM
Sequence: MAEVEAVQLKEEGNRHFQLQDYKAATNSYSQALKLTKDKALLATLYRNRAACGLKTESYVQAASDASRAIDINSSDIKALYRRCQALEHLGKLDQAFKDVQRCATLEPRNQNFQEMLRRLNTSIQEKLRVQFSTDSRVQKMFEILLDENSEADKREKAANNLIVLGREEAGAEKIFQNNGVALLLQLLDTKKPELVLAAVRTLSGMCSGHQARATVILHAVRIDRICSLMAVENEEMSLAVCNLLQAIIDSLSGEDKREHRGKEEALVLDTKKDLKQITSHLLDMLVSKKVSGQGRDQALNLLNKNVPRKDLAIHDNSRTIYVVDNGLRKILKVVGQVPDLPSCLPLTDNTRMLASILINKLYDDLRCDPERDHFRKICEEYITGKFDPQDMDKNLNAIQTVSGILQGPFDLGNQLLGLKGVMEMMVALCGSERETDQLVAVEALIHASTKLSRATFIITNGVSLLKQIYKTTKNEKIKIRTLVTSDKTILYSVATTLVNCTNSYDVKEVIPELVQLAKFSKQHVPEEHPKDKKDFIDMRVKRLLKAGVISALACMVKADSAILTDQTKELLARVFLALCDNPKDRGTIVAQGGGKALIPLALEGTDVGKVKAAHALAKIAAVSNPDIAFPGERVYEVVRPLVRLLDTQRDGLQNYEALLGLTNLSGRSDKLRQKIFKERALPDIENYMFENHDQLRQAATECMCNMVLHKEVQERFLADGNDRLKLVVLLCGEDDDKVQNAAAGALAMLTAAHKKLCLKMTQVTTQWLEILQRLCLHDQLSVQHRGLVIAYNLLAADAELAKKLVESELLEILTVVGKQEPDEKKAEVVQTARECLIKCMDYGFIKPVSSpecifications
unc-45 homolog B (C. elegans) | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human UNC45B protein. | |
UNC45B | |
CMYA4/FLJ38610/MGC119540/MGC119541/SMUNC45/UNC45 | |
Mouse | |
Affinity Purified | |
RUO | |
146862 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC101062 | |
UNC45B | |
UNC45B (AAI01063.1, 1 a.a. ∼ 850 a.a) full-length human protein. | |
50 μg | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction