Learn More
Uncoupling Protein 1 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Uncoupling Protein 1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in PBS with 0.1% BSA as stabilizer and 0.05% sodium azide. |
| Gene Symbols | UCP1; Thermogenin; UCP; SLC25A7 |
| Host Species | Rabbit |
| Immunogen | A 12 aa peptide at the C-terminus of the mouse/rat UCP1 (Bouillaud et al. 1986; Ridley et al. 1986; Kozak et al. 1988; Cassard et al. 1990; Miroux et al. 1993). |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.