Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Uncoupling Protein-2 (UCP-2) (144-157), Polyclonal

Catalog No. 662047100UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
66-204-7100UL 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66-204-7100UL Supplier MilliporeSigma™ Supplier No. 662047100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Uncoupling Protein-2 (UCP-2) (144-157) Clone: in Human, Mouse, Rat samples, and it is validated for ELISA, Immunocytochemistry, Immuno Blotting

The uncoupling protein is essential for the thermogenic properties of brown fat in mammals. It is inserted in the inner mitochondrial membrane by means of sixα-helical hydrophobic transmembrane domains.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Uncoupling Protein-2 (UCP-2) (144-157)
Applications ELISA, Immunoblot, Immunoblot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation In 140mM NaCl, 100mM potassium phosphate, pH 7.0.
Host Species Rabbit
Immunogen a synthetic peptide [(C)QARAGGGRRYQSTV] corresponding to amino acids 144-157 of human and mouse UCP-2
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the UCP-2 protein.
Target Species Human, Mouse, Rat
Content And Storage ≤ -70°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.