Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

unmethylated Histone H3 (Lys9) Mouse anti-Human, AlexaFluor™647, Clone: 9B1-2G6, MilliporeSigma™

Catalog No. MABE263AF64 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE263AF64 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE263AF64 Supplier MilliporeSigma Supplier No. MABE263AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-unmethylated Histone H3 (Lys9) Antibody, clone 9B1-2G6 is validated for use in Immunocytochemistry for the detection of unmethylated Histone H3 (Lys9).
TRUSTED_SUSTAINABILITY

Specifications

Antigen unmethylated Histone H3 (Lys9)
Applications Immunocytochemistry
Classification Monoclonal
Clone 9B1-2G6
Conjugate Alexa Fluor 647
Formulation Purified conjugated mouse monoclonal antibody in buffer containing PBS and 15mg/mL BSA with 0.1% Sodium Azide.
Gene Accession No. NP_038578
Gene Symbols HIST1H3A, H3FA, HIST1H3B, H3FL, HIST1H3C, H3FC, HIST1H3D, H3FB, HIST1H3E, H3FD, HIST1H3F, H3FI, HIST1H3G, H3FH, HIST1H3H, H3FK, HIST1H3I, H3FF, HIST1H3J, H3FJ
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human unmethylated Histone H3 (Lys9).
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.