Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-uPA Clone: 16,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ IM15L100UG
Description
Specifically detects uPA Clone: 16 in Human samples, and it is validated for Immunoprecipitation
Urokinase-type plasminogen activator (uPA) is a serine proteinase that functions in the conversion of plasminogen to the active proteinase, plasmin1. It is secreted from cells as an inactive 52kDa precursor (single-chain or sc-uPA) which is activated to the two-chain form consisting of an A and B-chain. The active high molecular weight form (HMW-uPA) can be further processed by removal of an amino-terminal fragment (ATF) to an active low molecular weight form (LMW-uPA) of ∽33kDa. The B-chain contains the active site and binds the two endogenous plasminogen activator inhibitors, PAI-1 and PAI-22. The binding of pro-uPA to its cell surface receptor enhances its rate of activation3. uPA is responsible for normal physiological processes such as tissue remodeling and cell migration. Since cell surface plasminogen activation is involved in extracellular matrix degradation, uPA is likely to play a key role in the process of invasion and metastasis.
Specifications
uPA | |
Monoclonal | |
Unconjugated | |
Mouse | |
100 μg | |
Primary | |
Human | |
Purified |
Immunoprecipitation | |
16 | |
Lyophilized from 20mM NH4HCO3, 100μg BSA. | |
the B chain of human urokinase plasminogen activator | |
RUO | |
Recognizes uPA/PAI complexes in dexamethasone-treated HT-1080 cells and carcinoma tissue. Sold under license of U.S. Patent 5,422,245. | |
2°C to 8°C | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction