Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-uPAR, PE, Clone: 109801, R&D Systems™
Supplier: R&D Systems FAB531P
This item is not returnable.
View return policy
Description
Applications:
Flow Cytometry
Specifications
uPAR | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, PLAUR, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5329 | |
Mouse | |
Purified |
Flow Cytometry | |
109801 | |
Flow Cytometry 10 uL/10^6 cells | |
P35456 | |
PLAUR | |
Mouse myeloma cell line NS0-derived recombinant mouse uPAR isoform 1 Leu24-Thr297 Accession # P35456 | |
100 Tests | |
Primary | |
Detects mouse uPAR in Western blots. In Western blots, this antibody does not cross-react with recombinant human uPAR. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction