Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UPP1, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89203406 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-203-406 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-203-406 Supplier Abnova Corporation Supplier No. H00007378K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human UPP1 peptide using ARM Technology.

The 2 known types of pyrimidine nucleoside phosphorylases, uridine phosphorylase (UP; EC 2.4.2.3) and thymidine phosphorylase (TP; EC 2.4.2.4), in the presence of orthophosphate, catalyze the reversible phosphorolysis of uridine and thymidine or deoxyuridine, respectively, to free bases and ribose-1-phosphate or deoxyribose-1-phosphate. Pyrimidine nucleoside phosphorylases can add ribose or deoxyribose to pyrimidine bases to form nucleosides that can be incorporated into RNA or DNA (Watanabe and Uchida, 1995 [PubMed 7488099]).[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen UPP1
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description uridine phosphorylase 1
Formulation In 1X PBS, pH 7.4
Gene UPP1
Gene Accession No. UPP1
Gene Alias UDRPASE, UP, UPASE, UPP
Gene Symbols UPP1
Host Species Rabbit
Immunogen A synthetic peptide of human UPP1
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7378
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.